This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00442722_g1
See other C2 GE Assays ›
Gene Symbol:
C2
Gene Name:
complement component 2 (within H-2S)
Gene Aliases:
-
Chromosome Location:
Chr.17: 34862602 - 34882100 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00442722_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013484.2 NP_038512.2 4 - 5 741 80
GenBank mRNA AK146812.1 - 4 - 5 742 80
AK213720.1 - 1 - 2 53 80
BC011086.1 - 4 - 5 704 3992881 80
M57891.1 - 4 - 5 688 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002817.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene C2

Gene Ontology Categories:

Function(s) Process(es)

immune system process
proteolysis
complement activation
complement activation, classical pathway
innate immune response
positive regulation of apoptotic cell clearance
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700018268.mm.1 NM_013484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018268.mm.1 NM_013484.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700018268.mm.1 TC1700002817.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700018267.mm.1 NM_013484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018267.mm.1 NM_013484.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700018267.mm.1 TC1700002817.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0