This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00441798_g1
See other TIE1 GE Assays ›
Gene Symbol:
Tie1
Gene Name:
tyrosine kinase with immunoglobulin-like and EGF-like domains 1
Gene Aliases:
D430008P04Rik, TIE, tie-1
Chromosome Location:
Chr.4: 118471191 - 118489849 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00441798_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tie1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011587.2 NP_035717.2 13 - 14 2352 85
GenBank mRNA AK051512.1 - 13 - 14 2298 85
AK052192.1 - 13 - 14 2272 85
AK052413.1 - 13 - 14 2285 85
AK161898.1 - 13 - 14 2271 85
BC046452.1 - 13 - 14 2274 4195879 85
BC057004.1 - 12 - 13 2718 6831929 85
BC060182.1 - 12 - 13 2718 6831929 85
X71425.1 - 13 - 14 2253 85
X73960.1 - 13 - 14 2252 85
X80764.1 - 13 - 14 2566 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003393.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tie1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
blood vessel development
vasculogenesis
in utero embryonic development
protein phosphorylation
phosphorylation
negative regulation of angiogenesis
negative regulation of cell migration
response to retinoic acid
plasma membrane fusion
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0