This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00441529_g1
See other SLPI GE Assays ›
Gene Symbol:
Slpi
Gene Name:
secretory leukocyte peptidase inhibitor
Gene Aliases:
-
Chromosome Location:
Chr.2: 164354070 - 164356507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00441529_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slpi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011414.3 NP_035544.1 2 - 3 485 93
GenBank mRNA AK010170.1 - 2 - 3 272 93
BC028509.1 - 2 - 3 485 1513866 93
U73004.1 - 696 93
U88093.1 - 2 - 3 262 93
U94341.1 - 2 - 3 263 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005112.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slpi

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of peptidase activity
negative regulation of endopeptidase activity
negative regulation of protein binding
negative regulation of viral genome replication
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
enzyme binding
peptidase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200043586.mm.1 NM_011414.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043586.mm.1 NM_011414.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200043586.mm.1 TC0200005112.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0