This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00441501_g1
See other SLC4A1 GE Assays ›
Gene Symbol:
Slc4a1
Gene Name:
solute carrier family 4 (anion exchanger), member 1
Gene Aliases:
Ae1, CD233, Empb3, l11Jus51
Chromosome Location:
Chr.11: 102348820 - 102365281 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00441501_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011403.2 NP_035533.1 12 - 13 1785 69
GenBank mRNA AK137542.1 - 12 - 13 1681 69
AK157181.1 - 12 - 13 1754 69
AK157833.1 - 12 - 13 1643 69
AK157861.1 - 12 - 13 1655 69
AK158143.1 - 12 - 13 1786 69
AK158184.1 - 12 - 13 1751 69
AK158243.1 - 12 - 13 1655 69
AK158453.1 - 12 - 13 1630 69
AK162616.1 - 12 - 13 1630 69
AK165195.1 - 12 - 13 1642 69
AK165267.1 - 12 - 13 1649 69
AK171662.1 - 1681 69
AK171808.1 - 12 - 13 1619 69
AK171845.1 - 1681 69
AK172103.1 - 12 - 13 1642 69
AK172316.1 - 12 - 13 1630 69
AK172395.1 - 12 - 13 1681 69
AK172456.1 - 12 - 13 1665 69
AK172569.1 - 12 - 13 1630 69
AY296129.1 - 12 - 13 1618 69
BC052419.1 - 12 - 13 1625 5702599 69
BC053429.1 - 12 - 13 1616 6506270 69
M29379.1 - 12 - 13 1612 69
X02677.1 - 12 - 13 1618 69
X03917.1 - 10 - 11 1462 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003833.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
chloride transport
inorganic anion transport
bicarbonate transport
regulation of intracellular pH
anion transmembrane transport
chloride transmembrane transport
actin binding
transporter activity
inorganic anion exchanger activity
protein binding
protein C-terminus binding
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
anion:anion antiporter activity
enzyme binding
ankyrin binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100035662.mm.1 NM_011403.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100035662.mm.1 NM_011403.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100035662.mm.1 TC1100003833.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0