This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00441493_m1
See other SLC4A1 GE Assays ›
Gene Symbol:
Slc4a1
Gene Name:
solute carrier family 4 (anion exchanger), member 1
Gene Aliases:
Ae1, CD233, Empb3, l11Jus51
Chromosome Location:
Chr.11: 102348820 - 102365281 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00441493_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011403.2 NP_035533.1 4 - 5 500 83
GenBank mRNA AK010967.1 - 4 - 5 329 83
AK137542.1 - 4 - 5 396 83
AK157181.1 - 4 - 5 469 83
AK157833.1 - 4 - 5 358 83
AK157861.1 - 4 - 5 370 83
AK158143.1 - 4 - 5 501 83
AK158184.1 - 4 - 5 466 83
AK158243.1 - 4 - 5 370 83
AK158453.1 - 4 - 5 345 83
AK162616.1 - 4 - 5 345 83
AK165195.1 - 4 - 5 357 83
AK165267.1 - 4 - 5 357 83
AK171662.1 - 396 83
AK171808.1 - 4 - 5 334 83
AK171845.1 - 396 83
AK172103.1 - 4 - 5 359 83
AK172316.1 - 4 - 5 345 83
AK172395.1 - 4 - 5 396 83
AK172456.1 - 4 - 5 382 83
AK172569.1 - 4 - 5 345 83
AY296129.1 - 4 - 5 333 83
BC052419.1 - 4 - 5 340 5702599 83
BC053429.1 - 4 - 5 331 6506270 83
M29379.1 - 4 - 5 327 83
X02677.1 - 4 - 5 333 83
X03917.1 - 2 - 3 177 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003833.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK165267.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
chloride transport
inorganic anion transport
bicarbonate transport
regulation of intracellular pH
anion transmembrane transport
chloride transmembrane transport
actin binding
transporter activity
inorganic anion exchanger activity
protein binding
protein C-terminus binding
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
anion:anion antiporter activity
enzyme binding
ankyrin binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100035671.mm.1 NM_011403.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100035671.mm.1 NM_011403.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100035671.mm.1 TC1100003833.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0