This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440840_m1
See other PRKACB GE Assays ›
Gene Symbol:
Prkacb
Gene Name:
protein kinase, cAMP dependent, catalytic, beta
Gene Aliases:
Pkacb
Chromosome Location:
Chr.3: 146729579 - 146812946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440840_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkacb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164198.1 NP_001157670.1 5 - 6 678 139
NM_001164199.1 NP_001157671.1 5 - 6 551 139
NM_001164200.1 NP_001157672.1 5 - 6 643 139
NM_011100.4 NP_035230.1 5 - 6 682 139
GenBank mRNA AK048319.1 - 5 - 6 674 139
AK139419.1 - 5 - 6 423 139
AK148728.1 - 5 - 6 492 139
AK150122.1 - 5 - 6 644 139
AK150134.1 - 5 - 6 644 139
AK163585.1 - 5 - 6 456 139
BC054533.1 - 5 - 6 666 5704796 139
J02626.1 - 5 - 6 611 139
JN189786.1 - 5 - 6 494 139
JN189787.1 - 5 - 6 443 139
JN189788.1 - 5 - 6 418 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003106.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Mouse Estrogen Panel Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkacb

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
protein phosphorylation
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
phosphorylation
negative regulation of meiotic cell cycle
regulation of protein processing
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_001164198.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_001164198.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_001164199.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_001164199.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_001164200.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_001164200.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_011100.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 NM_011100.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300024449.mm.1 TC0300003106.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 781049 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0