This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440692_m1
See other PDGFRA GE Assays ›
Gene Symbol:
Pdgfra
Gene Name:
platelet derived growth factor receptor, alpha polypeptide
Gene Aliases:
AI115593, CD140a, Pdgfr-2
Chromosome Location:
Chr.5: 75151401 - 75198211 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440692_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdgfra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083316.1 NP_001076785.1 13 - 14 2093 66
NM_011058.2 NP_035188.2 13 - 14 2076 66
GenBank mRNA AK035501.1 - 13 - 14 2034 66
AK081664.1 - 13 - 14 2075 66
AK136490.1 - 13 - 14 2062 66
AK145097.1 - 13 - 14 2677 66
AK147267.1 - 13 - 14 2025 66
AK163952.1 - 13 - 14 2140 66
BC053036.1 - 13 - 14 2093 5704645 66
M57683.1 - 13 - 14 1975 66
M84607.1 - 13 - 14 2086 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000709.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdgfra

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

luteinization
in utero embryonic development
hematopoietic progenitor cell differentiation
protein phosphorylation
chemotaxis
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cytosolic calcium ion concentration
multicellular organism development
estrogen metabolic process
positive regulation of cell proliferation
female gonad development
anatomical structure morphogenesis
organ morphogenesis
negative regulation of platelet activation
positive regulation of phospholipase C activity
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
signal transduction involved in regulation of gene expression
extracellular matrix organization
lung development
adrenal gland development
positive regulation of cell migration
male genitalia development
Leydig cell differentiation
cellular response to reactive oxygen species
platelet-derived growth factor receptor-alpha signaling pathway
cellular response to platelet-derived growth factor stimulus
positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
wound healing
odontogenesis of dentin-containing tooth
positive regulation of phosphatidylinositol 3-kinase activity
positive regulation of DNA replication
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
embryonic digestive tract morphogenesis
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
regulation of chemotaxis
cardiac myofibril assembly
palate development
face morphogenesis
cell chemotaxis
retina vasculature development in camera-type eye
positive regulation of ERK1 and ERK2 cascade
platelet aggregation
cellular response to amino acid stimulus
metanephric glomerular capillary formation
regulation of mesenchymal stem cell differentiation
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
platelet-derived growth factor alpha-receptor activity
vascular endothelial growth factor-activated receptor activity
platelet-derived growth factor receptor binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein complex binding
vascular endothelial growth factor binding
protein homodimerization activity
phosphatidylinositol 3-kinase binding
platelet-derived growth factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500006266.mm.1 NM_001083316.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500006266.mm.1 NM_011058.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500006266.mm.1 TC0500000709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500006267.mm.1 NM_001083316.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500006267.mm.1 NM_011058.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500006267.mm.1 TC0500000709.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0