This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440554_m1
See other GPR143 GE Assays ›
Gene Symbol:
Gpr143
Gene Name:
G protein-coupled receptor 143
Gene Aliases:
Oa1
Chromosome Location:
Chr.X: 152781921 - 152808651 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440554_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010951.3 NP_035081.3 4 - 5 669 101
GenBank mRNA AK185813.1 - 1 - 2 102 101
BC119384.1 - 4 - 5 585 8734133 101
BC137813.1 - 4 - 5 585 8860832 101
CJ131873.1 - 3 - 4 575 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001549.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr143

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
chemical synaptic transmission
melanosome localization
melanosome transport
melanosome organization
calcium-mediated signaling using intracellular calcium source
phosphatidylinositol-mediated signaling
regulation of calcium-mediated signaling
signal transducer activity
G-protein coupled receptor activity
dopamine binding
L-DOPA receptor activity
L-DOPA binding
tyrosine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00009864.mm.1 NM_010951.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00009864.mm.1 NM_010951.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00009864.mm.1 TC0X00001549.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0