This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440448_m1
See other NDRG1 GE Assays ›
Gene Symbol:
Ndrg1
Gene Name:
N-myc downstream regulated gene 1
Gene Aliases:
CAP43, CMT4D, DRG1, HMSNL, NMSL, Ndr1, Ndrl, PROXY1, RTP, TDD5
Chromosome Location:
Chr.15: 66929318 - 66969641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440448_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndrg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008681.2 NP_032707.2 4 - 5 361 77
GenBank mRNA AK004548.1 - 4 - 5 361 77
AK098075.1 - 4 - 5 360 77
AK161860.1 - 4 - 5 363 77
AK165007.1 - 4 - 5 361 77
AK170281.1 - 4 - 5 362 77
AK170439.1 - 4 - 5 360 77
AK170636.1 - 4 - 5 360 77
AK170809.1 - 4 - 5 361 77
AW611038.1 - 4 - 5 361 77
BC015282.1 - 4 - 5 316 4237943 77
BC071235.1 - 4 - 5 369 4242075 77
BC141246.1 - 4 - 5 252 9055858 77
BC171968.1 - 4 - 5 252 9054254 77
CA320478.1 - 1 - 2 104 77
U52073.1 - 4 - 5 340 77
U60593.1 - 4 - 5 359 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndrg1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
peripheral nervous system myelin maintenance
mast cell activation
positive regulation of spindle checkpoint
microtubule binding
Rab GTPase binding
gamma-tubulin binding
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0