This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440283_g1
See other MEA1 GE Assays ›
Gene Symbol:
Mea1
Gene Name:
male enhanced antigen 1
Gene Aliases:
Mea-1
Chromosome Location:
Chr.17: 46679138 - 46683126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440283_g1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mea1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277309.1 NP_001264238.1 2 - 3 425 67
NM_001277310.1 NP_001264239.1 2 - 3 327 67
NM_001277311.1 NP_001264240.1 2 - 3 343 67
NM_001277312.1 NP_001264241.1 2 - 3 370 67
NM_010787.2 NP_034917.1 2 - 3 362 67
NR_102392.1 2 - 3 454 67
GenBank mRNA AK005106.1 - 2 - 3 311 67
AK005369.1 - 2 - 3 315 67
AK008194.1 - 2 - 3 321 67
AK008305.1 - 2 - 3 323 67
AK010863.1 - 2 - 3 332 67
AK010904.1 - 2 - 3 360 67
AK010962.1 - 2 - 3 317 67
AK011189.1 - 2 - 3 304 67
AK012617.1 - 2 - 3 317 67
BC013344.1 - 1 - 2 325 4035701 67
BG085669.2 - 2 - 3 322 67
BY707372.1 - 2 - 3 408 67
CJ042312.1 - 2 - 3 413 67
D17341.1 - 2 - 3 301 67
L10401.1 - 2 - 3 346 67
M27938.1 - 2 - 3 346 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000900.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mea1

Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
cell differentiation

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208332 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 L10401.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 M27938.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 AK010863.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 AK010904.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 AK010962.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 L10401.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 M27938.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 AK010863.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 AK010904.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448489 AK010962.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892874 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 L10401.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 M27938.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 AK010863.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 AK010904.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 AK010962.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 L10401.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 M27938.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 AK010863.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 AK010904.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448489 AK010962.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208334 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208343 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208339 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208333 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208337 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208341 - -
Whole Mouse Genome G4122A A_51_P448489 L10401.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 M27938.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 AK010863.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 AK010904.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 AK010962.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 L10401.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 M27938.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 AK010863.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 AK010904.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P448489 AK010962.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P271031 L10401.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P271031 M27938.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P271031 AK010863.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P271031 AK010904.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P271031 AK010962.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139868 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13229823 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13229809 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31208332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208339 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31208335 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31208334 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13472102 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13229806 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31208341 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31208337 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31208333 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31208343 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10445605 - -
Affymetrix Mouse Gene 1.0 ST Array 10445606 - -
Affymetrix Mouse Gene 1.0 ST Array 10445603 - -
Affymetrix Mouse Gene 1.0 ST Array 10445604 - -
Affymetrix HT MG-430 PM Array Plate 1416810_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1437943_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10445601 - -
Mouse Genome 430 2.0 Array 1437943_PM_s_at - -
Mouse Genome 430 2.0 Array 1416810_at - -
Mouse Genome 430 2.0 Array 1437943_s_at - -
Mouse Genome 430 2.0 Array 1416810_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 17256_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 17256_PM_at - -
Affymetrix Mouse Clariom S Array PSR1700008619.mm.1 NM_001277312.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008619.mm.1 TC1700000900.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700008612.mm.1 NM_001277311.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008612.mm.1 TC1700000900.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700008618.mm.1 NM_001277312.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008618.mm.1 NM_010787.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700008618.mm.1 TC1700000900.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 NM_001277309.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 NM_001277310.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 NM_001277311.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 NM_001277312.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 NM_010787.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 NR_102392.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008622.mm.1 TC1700000900.mm.1 MTA TC
Mouse Expression Set 430 1416810_at - -
Mouse Expression Set 430 1437943_PM_s_at - -
Mouse Expression Set 430 1416810_PM_at - -
Mouse Expression Set 430 1437943_s_at - -
Murine Genome U74v2 Set 94891_PM_s_at - -
Murine Genome U74v2 Set 94891_s_at - -
Murine Genome U74v2 Set 94892_r_at - -
Murine Genome U74v2 Set 94892_PM_r_at - -
Murine Genome U74v2 Set 94890_at - -
Murine Genome U74v2 Set 94890_PM_at - -
Murine Genome U74v2 Set 170151_PM_r_at - -
Murine Genome U74v2 Set 170151_r_at - -
Mouse Genome 430A 2.0 Array 1437943_s_at - -
Mouse Genome 430A 2.0 Array 1416810_PM_at - -
Mouse Genome 430A 2.0 Array 1437943_PM_s_at - -
Mouse Genome 430A 2.0 Array 1416810_at - -
Affymetrix HT Mouse Genome 430A Array 1416810_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1437943_s_at - -
Affymetrix HT Mouse Genome 430A Array 1416810_at - -
Affymetrix HT Mouse Genome 430A Array 1437943_PM_s_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 910886 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6350338 - -
Illumina mouse-6 v1.0 expression beadchip 2230347 - -
Illumina mouse-6 v1.0 expression beadchip 6350338 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002230347 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006350338 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2750258 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2765450 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766403 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2766403 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2750258 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2765450 - -
mouse-6 v1.1 (Illumina) scl017256.3_8-S - -
Illumina Sentrix mouse-6.v1 bead array 6350338 - -
Illumina Sentrix mouse-6.v1 bead array 2230347 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl017256.3_8-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230347 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350338 - -
MouseRef-8 v2 Expression BeadChip ILMN_2750258 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2765450 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2750258 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl017256.3_8-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0