This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440281_g1
See other MDK GE Assays ›
Gene Symbol:
Mdk
Gene Name:
midkine
Gene Aliases:
MK, Mek
Chromosome Location:
Chr.2: 91929805 - 91932297 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440281_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mdk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012335.2 NP_001012335.1 3 - 4 405 81
NM_001012336.2 NP_001012336.1 3 - 4 402 81
NM_001291481.1 NP_001278410.1 3 - 4 312 81
NM_001291482.1 NP_001278411.1 2 - 3 325 81
NM_010784.5 NP_034914.1 3 - 4 604 81
GenBank mRNA AK032742.1 - 3 - 4 308 81
AK084457.1 - 3 - 4 325 81
BC012244.1 - 3 - 4 273 4167496 81
CF728526.1 - 3 - 4 298 81
CF731708.1 - 3 - 4 401 81
CF738400.1 - 3 - 4 410 81
CF740555.1 - 1 - 2 122 81
CJ174578.1 - 2 - 3 200 81
M19662.1 - 2 - 3 483 81
M34327.1 - 3 - 4 278 81
M34328.1 - 3 - 4 589 81
M35833.1 - 3 - 4 278 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004129.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mdk

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
multicellular organism development
short-term memory
response to wounding
cell migration
cerebellar granular layer development
hippocampus development
cell differentiation
adrenal gland development
defecation
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
regulation of behavior
positive regulation of cell division
negative regulation of neuron death
growth factor activity
heparin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200034583.mm.1 NM_001012335.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034583.mm.1 NM_001012336.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034583.mm.1 NM_010784.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200034583.mm.1 TC0200004129.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0