Assay Details
Gene Symbol: Il1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010555.4 | NP_034685.1 | 1 - 2 | 47 | 78 | |
GenBank mRNA | AK132264.1 | - | 1 - 2 | 74 | 78 | |
X59769.1 | - | 1 - 2 | 47 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK132264.1,NM_010555.4,X59769.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:

Citations (1)
Accelerated recovery from acute hypoxia in obese mice is due to obesity-associated up-regulation of interleukin-1 receptor antagonist.
- Authors
- Sherry CL, Kim SS, Freund GG
- Journal
- Endocrinology - Page No 2660 (Mon Jun 01 00:00:00 UTC 2009)
- Volume (Year)
- 150 (2009)
- Product Usage
- -
