This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00438919_m1
See other FGF5 GE Assays ›
Gene Symbol:
Fgf5
Gene Name:
fibroblast growth factor 5
Gene Aliases:
Fgf-5, HBGF-5, angora, go
Chromosome Location:
Chr.5: 98254184 - 98277033 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00438919_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010203.5 NP_034333.1 2 - 3 687 135
GenBank mRNA AK028694.1 - 2 - 3 544 135
AK028894.1 - 2 - 3 689 135
BC071227.1 - 2 - 3 660 30443152 135
M30643.1 - 2 - 3 459 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000948.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgf5

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
glial cell differentiation
signal transduction involved in regulation of gene expression
positive regulation of cell division
receptor binding
fibroblast growth factor receptor binding
growth factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500008156.mm.1 NM_001277268.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500008156.mm.1 NM_010203.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500008156.mm.1 TC0500000948.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0