Assay Details
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010167.4 | NP_034297.2 | 8 - 9 | 863 | 106 | |
GenBank mRNA | AK041625.1 | - | 8 - 9 | 864 | 106 | |
BC120898.1 | - | 8 - 9 | 847 | 40061597 | 106 | |
BC120899.1 | - | 8 - 9 | 847 | 40061599 | 106 | |
Y17115.1 | - | 9 - 10 | 974 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
