This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00438814_m1
See other EYA3 GE Assays ›
Gene Symbol:
Eya3
Gene Name:
eyes absent 3 homolog (Drosophila)
Gene Aliases:
AI844637
Chromosome Location:
Chr.4: 132639016 - 132724765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00438814_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010166.3 NP_034296.2 12 - 13 1153 105
NM_210071.2 NP_997592.1 11 - 12 1114 105
NM_211357.2 NP_997596.1 9 - 10 933 105
GenBank mRNA AK144653.1 - 11 - 12 1105 105
BC052860.1 - 10 - 11 917 105
BC063259.1 - 11 - 12 1042 6406090 105
U61112.1 - 8 - 9 818 105
U81604.1 - 12 - 13 1121 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001594.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya3

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
protein dephosphorylation
cellular response to DNA damage stimulus
multicellular organism development
response to ionizing radiation
chromatin modification
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
positive regulation of DNA repair
chromatin binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400012524.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012524.mm.1 NM_210071.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012524.mm.1 NM_211357.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012524.mm.1 TC0400001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400012526.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012526.mm.1 NM_210071.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012526.mm.1 NM_211357.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012526.mm.1 TC0400001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400012527.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012527.mm.1 NM_210071.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012527.mm.1 NM_211357.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012527.mm.1 TC0400001594.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header