This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00438703_m1
See other EIF2AK3 GE Assays ›
Gene Symbol:
Eif2ak3
Gene Name:
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases:
AI427929, Pek, Perk
Chromosome Location:
Chr.6: 70844484 - 70905240 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00438703_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2ak3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010121.2 NP_034251.2 7 - 8 1486 64
GenBank mRNA AF076681.1 - 7 - 8 1478 64
AK139679.1 - 5 - 6 808 64
BC054809.1 - 7 - 8 1498 6839221 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000813.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2ak3

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
ossification
angiogenesis
chondrocyte development
translation
regulation of translation
protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
ER overload response
response to unfolded protein
endoplasmic reticulum organization
lactation
positive regulation of signal transduction
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of gene expression
regulation of translational initiation by eIF2 alpha phosphorylation
phosphorylation
negative regulation of translation
peptidyl-serine phosphorylation
regulation of fatty acid metabolic process
calcium-mediated signaling
bone mineralization
endoplasmic reticulum unfolded protein response
pancreas development
endocrine pancreas development
negative regulation of myelination
negative regulation of translation in response to stress
positive regulation of protein binding
SREBP signaling pathway
response to endoplasmic reticulum stress
PERK-mediated unfolded protein response
cellular response to glucose starvation
negative regulation of apoptotic process
fat cell differentiation
positive regulation of transcription from RNA polymerase I promoter
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
protein homooligomerization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
positive regulation of protein localization to nucleus
negative regulation of translation in response to endoplasmic reticulum stress
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of glutathione biosynthetic process
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
enzyme binding
protein phosphatase binding
identical protein binding
Hsp90 protein binding
eukaryotic initiation factor eIF2 binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600006194.mm.1 NM_010121.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600006194.mm.1 TC0600000813.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0