This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00438681_m1
See other EGF GE Assays ›
Gene Symbol:
Egf
Gene Name:
epidermal growth factor
Gene Aliases:
AI790464
Chromosome Location:
Chr.3: 129677569 - 129755322 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00438681_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010113.3 NP_034243.2 8 - 9 1682 62
GenBank mRNA AK085707.1 - 8 - 9 1249 62
AK088408.1 - 3 - 4 269 62
AK136439.1 - 3 - 4 275 62
AK142666.1 - 8 - 9 1378 62
AK157778.1 - 3 - 4 274 62
BC017681.1 - 9 - 10 1619 4221592 62
BC060741.1 - 8 - 9 1667 4164289 62
BC092277.1 - 8 - 9 1667 4240429 62
J00380.1 - 8 - 9 1677 62
V00741.1 - 8 - 9 1633 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002932.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK136439.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Egf

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
angiogenesis
epidermal growth factor receptor signaling pathway
STAT protein import into nucleus
positive regulation of cell proliferation
positive regulation of peptidyl-threonine phosphorylation
peptidyl-tyrosine phosphorylation
positive regulation of cerebellar granule cell precursor proliferation
positive regulation of catenin import into nucleus
positive regulation of phosphorylation
positive regulation of DNA binding
positive regulation of MAP kinase activity
positive regulation of epidermal growth factor-activated receptor activity
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
branching morphogenesis of an epithelial tube
regulation of protein secretion
regulation of peptidyl-tyrosine phosphorylation
negative regulation of secretion
regulation of protein transport
mammary gland alveolus development
ERK1 and ERK2 cascade
regulation of calcium ion import
negative regulation of cholesterol efflux
positive regulation of hyaluronan biosynthetic process
regulation of protein localization to cell surface
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
epidermal growth factor receptor binding
calcium ion binding
protein binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0