This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437116_m1
See other TNNC2 GE Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C2, fast
Gene Aliases:
Tncs
Chromosome Location:
Chr.2: 164777161 - 164779734 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437116_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnnc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009394.2 NP_033420.1 1 - 2 75 116
GenBank mRNA AK133548.1 - 1 - 2 71 116
BC024390.1 - 1 - 2 74 5026329 116
CB566378.1 - 110 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005130.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK133548.1,BC024390.1,NM_009394.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnnc2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

skeletal muscle contraction
actin binding
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200043740.mm.1 NM_009394.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043740.mm.1 NM_009394.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200043740.mm.1 TC0200005130.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200043739.mm.1 NM_009394.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043739.mm.1 NM_009394.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200043739.mm.1 TC0200005130.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0