Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500001642.mm.1 |
NM_001253892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001642.mm.1 |
NM_001253892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001642.mm.1 |
NM_009207.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001642.mm.1 |
NM_009207.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001642.mm.1 |
TC0500000215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500001643.mm.1 |
NM_001253892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001643.mm.1 |
NM_001253892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001643.mm.1 |
NM_009207.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001643.mm.1 |
NM_009207.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500001643.mm.1 |
TC0500000215.mm.1 |
MTA TC |