This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00436621_g1
See other SLC4A2 GE Assays ›
Gene Symbol:
Slc4a2
Gene Name:
solute carrier family 4 (anion exchanger), member 2
Gene Aliases:
Ae2, B3RP
Chromosome Location:
Chr.5: 24423761 - 24440947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00436621_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253892.1 NP_001240821.1 5 - 6 883 57
NM_009207.3 NP_033233.2 6 - 7 1079 57
GenBank mRNA AK145930.1 - 6 - 7 1004 57
AK147858.1 - 6 - 7 1035 57
AK180093.1 - 2 - 3 360 57
AK189722.1 - 1 - 2 169 57
AK213497.1 - 1 - 2 93 57
AK219021.1 - 2 - 3 320 57
BC054102.1 - 5 - 6 888 30017310 57
J04036.1 - 6 - 7 994 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000215.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
chloride transport
bicarbonate transport
regulation of intracellular pH
chloride transmembrane transport
transporter activity
inorganic anion exchanger activity
anion transmembrane transporter activity
chloride transmembrane transporter activity
antiporter activity
anion:anion antiporter activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500001624.mm.1 NM_001253892.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001624.mm.1 NM_001253892.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001624.mm.1 NM_009207.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001624.mm.1 NM_009207.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500001624.mm.1 TC0500000215.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500001625.mm.1 NM_001253892.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001625.mm.1 NM_001253892.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001625.mm.1 NM_009207.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001625.mm.1 NM_009207.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500001625.mm.1 TC0500000215.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0