This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00436370_g1
See other MST1R GE Assays ›
Gene Symbol:
Mst1r
Gene Name:
macrophage stimulating 1 receptor (c-met-related tyrosine kinase)
Gene Aliases:
CD136, CDw136, Fv-2, Fv2, PTK8, Ron, STK
Chromosome Location:
Chr.9: 107906860 - 107920383 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00436370_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mst1r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009074.2 NP_033100.2 6 - 7 2309 75
NR_109782.1 6 - 7 2337 75
GenBank mRNA AK053655.1 - 6 - 7 2320 75
BC141379.1 - 6 - 7 2120 9055992 75
X74736.1 - 6 - 7 2309 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001383.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mst1r

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

immune system process
protein phosphorylation
response to virus
viral process
phosphorylation
positive regulation of MAP kinase activity
innate immune response
positive regulation of protein kinase B signaling
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900010910.mm.1 NM_009074.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010910.mm.1 NM_009074.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900010910.mm.1 NR_109782.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900010910.mm.1 TC0900001383.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0