This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00436318_m1
See other RET GE Assays ›
Gene Symbol:
Ret
Gene Name:
ret proto-oncogene
Gene Aliases:
PTC, RET51, RET9, c-Ret
Chromosome Location:
Chr.6: 118151748 - 118197744 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00436318_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ret
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080780.1 NP_001074249.1 17 - 18 3553 73
NM_009050.2 NP_033076.2 17 - 18 3553 73
GenBank mRNA AF209436.1 - 17 - 18 2942 73
AK044686.1 - 17 - 18 3139 73
AK051633.1 - 17 - 18 3132 73
AK084977.1 - 17 - 18 3203 73
AY326397.1 - 17 - 18 2942 73
BC059012.1 - 17 - 18 3527 6849696 73
X67812.1 - 17 - 18 2942 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002996.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ret

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
ureteric bud development
neural crest cell migration
embryonic epithelial tube formation
protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
neuron cell-cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
nervous system development
anatomical structure morphogenesis
positive regulation of gene expression
positive regulation of neuron projection development
positive regulation of neuron maturation
phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron differentiation
positive regulation of cell migration
membrane protein proteolysis
positive regulation of cell adhesion mediated by integrin
ureter maturation
neuron maturation
positive regulation of cell size
positive regulation of transcription, DNA-templated
response to pain
enteric nervous system development
regulation of axonogenesis
innervation
Peyer's patch morphogenesis
cellular response to retinoic acid
positive regulation of metanephric glomerulus development
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
calcium ion binding
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600023018.mm.1 NM_001080780.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023018.mm.1 NM_001080780.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023018.mm.1 NM_009050.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023018.mm.1 NM_009050.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600023018.mm.1 TC0600002996.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0