This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435914_m1
See other PRKAR2A GE Assays ›
Gene Symbol:
Prkar2a
Gene Name:
protein kinase, cAMP dependent regulatory, type II alpha
Gene Aliases:
1110061A24Rik, AI317181, AI836829, RII(alpha)
Chromosome Location:
Chr.9: 108692143 - 108749512 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435914_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkar2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008924.2 NP_032950.1 1 - 2 566 84
GenBank mRNA AF533977.1 - 1 - 2 566 84
AK132728.1 - 1 - 2 505 84
BC075623.1 - 1 - 2 537 6854057 84
BC080276.1 - 1 - 2 560 30354403 84
J02935.1 - 1 - 2 249 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001419.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkar2a

Panther Classification:

Molecular Function -

protein-binding activity modulator kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
regulation of protein kinase activity
protein heterooligomerization
negative regulation of cAMP-dependent protein kinase activity
nucleotide binding
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
small molecule binding
protein homodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900011491.mm.1 NM_008924.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011491.mm.1 NM_008924.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011491.mm.1 TC0900001419.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900011490.mm.1 NM_008924.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011490.mm.1 NM_008924.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900011490.mm.1 TC0900001419.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 887925 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0