Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900000522.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000522.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000522.mm.1 |
TC0900000069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000521.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000521.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000521.mm.1 |
TC0900000069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000516.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000516.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000516.mm.1 |
TC0900000069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000518.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000518.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000518.mm.1 |
TC0900000069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000519.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000519.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000519.mm.1 |
TC0900000069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000520.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000520.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000520.mm.1 |
TC0900000069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000524.mm.1 |
NM_008829.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000524.mm.1 |
NM_008829.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000524.mm.1 |
TC0900000069.mm.1 |
MTA TC |