This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435424_m1
See other NTRK2 GE Assays ›
Gene Symbol:
Ntrk2
Gene Name:
neurotrophic tyrosine kinase, receptor, type 2
Gene Aliases:
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB
Chromosome Location:
Chr.13: 58806569 - 59133970 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435424_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ntrk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025074.2 NP_001020245.1 11 - 12 1954 74
NM_001282961.1 NP_001269890.1 11 - 12 1921 74
NM_008745.3 NP_032771.1 11 - 12 1921 74
GenBank mRNA AB377224.1 - 10 - 11 1331 74
AB377225.1 - 10 - 11 1331 74
AK009606.1 - 10 - 11 1308 74
AK018789.1 - 11 - 12 1852 74
AK028827.1 - 11 - 12 1774 74
AK147391.1 - 11 - 12 1752 74
AK147441.1 - 10 - 11 1742 74
AK160789.1 - 11 - 12 1922 74
BC052014.1 - 10 - 11 1716 5707891 74
M33385.1 - 10 - 11 1293 74
X17647.1 - 1804 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000718.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ntrk2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
neuron migration
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
nervous system development
neuromuscular junction development
learning
long-term memory
feeding behavior
positive regulation of cell proliferation
positive regulation of gene expression
positive regulation of neuron projection development
glutamate secretion
positive regulation of phosphatidylinositol 3-kinase signaling
phosphorylation
peptidyl-tyrosine phosphorylation
regulation of metabolic process
central nervous system neuron development
cerebral cortex development
cell differentiation
neuron differentiation
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
neurotrophin signaling pathway
mechanoreceptor differentiation
regulation of GTPase activity
regulation of MAPK cascade
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
retinal rod cell development
protein autophosphorylation
regulation of neurotransmitter secretion
oligodendrocyte differentiation
peripheral nervous system neuron development
positive regulation of axonogenesis
regulation of dendrite development
regulation of protein kinase B signaling
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
retina development in camera-type eye
long-term synaptic potentiation
cellular response to amino acid stimulus
positive regulation of glucocorticoid receptor signaling pathway
negative regulation of anoikis
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
neurotrophin receptor activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein homodimerization activity
neurotrophin binding
ephrin receptor binding
brain-derived neurotrophic factor binding
brain-derived neurotrophic factor-activated receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300004737.mm.1 NM_001025074.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004737.mm.1 NM_001282961.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004737.mm.1 NM_008745.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004737.mm.1 TC1300000718.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004743.mm.1 NM_001025074.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004743.mm.1 NM_001282961.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004743.mm.1 TC1300000718.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0