This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435320_g1
See other NPR1 GE Assays ›
Gene Symbol:
Npr1
Gene Name:
natriuretic peptide receptor 1
Gene Aliases:
AI893888, GC-A, NPR-A, NPRA, Pndr
Chromosome Location:
Chr.3: 90450592 - 90465928 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435320_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008727.5 NP_032753.5 16 - 17 2910 68
GenBank mRNA AK135008.1 - 7 - 8 853 68
BC110659.1 - 16 - 17 2790 30847317 68
BC139767.1 - 16 - 17 2769 68
J05504.1 - 17 - 18 2894 68
L31932.1 - 16 - 17 2558 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Npr1

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
protein phosphorylation
cell surface receptor signaling pathway
receptor guanylyl cyclase signaling pathway
regulation of blood pressure
cyclic nucleotide biosynthetic process
positive regulation of cGMP biosynthetic process
intracellular signal transduction
dopamine metabolic process
negative regulation of smooth muscle cell proliferation
regulation of blood vessel size
nucleotide binding
guanylate cyclase activity
protein kinase activity
ATP binding
GTP binding
transferase activity, transferring phosphorus-containing groups
lyase activity
phosphorus-oxygen lyase activity
natriuretic peptide receptor activity
peptide hormone binding
protein kinase binding
hormone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0