This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435166_m1
See other NNT GE Assays ›
Gene Symbol:
Nnt
Gene Name:
nicotinamide nucleotide transhydrogenase
Gene Aliases:
4930423F13Rik, AI323702, BB168308
Chromosome Location:
Chr.13: 119335527 - 119409138 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435166_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nnt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008710.3 NP_032736.2 2789 79
GenBank mRNA AK033282.1 - 12 - 13 1977 79
AK040460.1 - 10 - 11 1655 79
AK050478.1 - 11 - 12 2694 79
AK052234.1 - 12 - 13 2021 79
AK054332.1 - 12 - 13 2022 79
AK083413.1 - 12 - 13 2020 79
AK084546.1 - 12 - 13 2019 79
AK084604.1 - 12 - 13 2019 79
AK085903.1 - 10 - 11 1680 79
AK088044.1 - 12 - 13 2771 79
AK090115.1 - 2792 79
AK129006.1 - 12 - 13 2766 79
AK159694.1 - 13 - 15 2775 79
AK168736.1 - 11 - 12 2572 79
BC008518.1 - 12 - 13 2752 3601702 79
Z49204.1 - 11 - 12 2696 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nnt

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

proton transport
cell redox homeostasis
oxidation-reduction process
NAD(P)+ transhydrogenase activity
NAD(P)+ transhydrogenase (AB-specific) activity
oxidoreductase activity
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0