This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433882_g1
See other GUCY2E GE Assays ›
Gene Symbol:
Gucy2e
Gene Name:
guanylate cyclase 2e
Gene Aliases:
GC-E, GC1, ROS-GC1
Chromosome Location:
Chr.11: 69218117 - 69237358 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433882_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gucy2e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008192.3 NP_032218.2 17 - 18 3231 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gucy2e

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
protein phosphorylation
visual perception
cyclic nucleotide biosynthetic process
intracellular signal transduction
response to stimulus
nucleotide binding
guanylate cyclase activity
protein kinase activity
ATP binding
GTP binding
transferase activity, transferring phosphorus-containing groups
lyase activity
phosphorus-oxygen lyase activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0