This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433694_g1
See other GPR12 GE Assays ›
Gene Symbol:
Gpr12
Gene Name:
G-protein coupled receptor 12
Gene Aliases:
Gpcr01, Gpcr12, Gpcr20, Gpcr21
Chromosome Location:
Chr.5: 146582150 - 146586487 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433694_g1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA D21061.1 - 1 - 2 88 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the D21061.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0