This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433597_m1
See other GIF GE Assays ›
Gene Symbol:
Gif
Gene Name:
gastric intrinsic factor
Gene Aliases:
AV073125
Chromosome Location:
Chr.19: 11747543 - 11763447 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433597_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gif
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008118.3 NP_032144.2 6 - 7 962 132
GenBank mRNA AK078933.1 - 6 - 7 963 132
BC118519.1 - 6 - 7 948 40105765 132
L24191.1 - 6 - 7 874 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000231.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gif

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cobalt ion transport
cobalamin metabolic process
cobalamin transport
cobalamin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002811.mm.1 NM_008118.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002811.mm.1 NM_008118.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900002811.mm.1 TC1900000231.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header