This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433314_m1
See other FGFR4 GE Assays ›
Gene Symbol:
Fgfr4
Gene Name:
fibroblast growth factor receptor 4
Gene Aliases:
Fgfr-4
Chromosome Location:
Chr.13: 55152818 - 55168759 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433314_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgfr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008011.2 NP_032037.2 5 - 6 756 92
GenBank mRNA AK084850.1 - 5 - 6 756 92
AK085723.1 - 5 - 6 755 92
AY493377.2 - 4 - 5 642 92
BC033313.1 - 5 - 6 943 5132831 92
DQ388428.1 - 4 - 5 642 92
X59927.1 - 5 - 6 698 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000650.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Mouse Tumor Metastasis Panel Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgfr4

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

organ induction
protein phosphorylation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of gene expression
regulation of extracellular matrix disassembly
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
regulation of lipid metabolic process
lung development
glucose homeostasis
positive regulation of catalytic activity
positive regulation of proteolysis
protein autophosphorylation
phosphate ion homeostasis
alveolar secondary septum development
positive regulation of ERK1 and ERK2 cascade
regulation of bile acid biosynthetic process
regulation of cholesterol homeostasis
positive regulation of DNA biosynthetic process
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
protein binding
ATP binding
heparin binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
fibroblast growth factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300004254.mm.1 NM_008011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004254.mm.1 NM_008011.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004254.mm.1 TC1300000650.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004255.mm.1 NM_008011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004255.mm.1 NM_008011.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004255.mm.1 TC1300000650.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0