This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433284_m1
See other FGF18 GE Assays ›
Gene Symbol:
Fgf18
Gene Name:
fibroblast growth factor 18
Gene Aliases:
D130055P09Rik, FGF-18
Chromosome Location:
Chr.11: 33116970 - 33147400 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433284_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008005.2 NP_032031.1 2 - 3 200 89
GenBank mRNA AB004639.1 - 2 - 3 73 89
AF075291.1 - 2 - 3 208 89
AF211187.1 - 2 - 3 73 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002484.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgf18

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

ossification
angiogenesis
intramembranous ossification
endochondral ossification
chondrocyte development
signal transduction
nervous system development
cell proliferation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
lung development
positive regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of chondrocyte differentiation
positive regulation of MAP kinase activity
positive regulation of ERK1 and ERK2 cascade
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
receptor binding
fibroblast growth factor receptor binding
type 1 fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0