This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433132_m1
See other PDIA3 GE Assays ›
Gene Symbol:
Pdia3
Gene Name:
protein disulfide isomerase associated 3
Gene Aliases:
58kDa, ERp57, ERp60, ERp61, Erp, Grp58, PDI, PDI-Q2, PI-PLC, PLC[a], Plca
Chromosome Location:
Chr.2: 121413902 - 121438687 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433132_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdia3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007952.2 NP_031978.2 4 - 5 607 120
GenBank mRNA AK088721.1 - 4 - 5 567 120
AK132459.1 - 4 - 5 571 120
AK133799.1 - 4 - 5 566 120
AK167807.1 - 4 - 5 560 120
AK169611.1 - 4 - 5 561 120
BC003285.1 - 4 - 5 526 3582199 120
BC033439.1 - 4 - 5 501 4016310 120
BQ714617.1 - 607 120
M73329.1 - 4 - 5 546 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001812.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdia3

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
proteolysis
response to endoplasmic reticulum stress
cell redox homeostasis
oxidation-reduction process
cellular response to vitamin D
positive regulation of protein folding
positive regulation of extrinsic apoptotic signaling pathway
protein disulfide isomerase activity
protein binding
peptidase activity
isomerase activity
protein-disulfide reductase (glutathione) activity
MHC class I protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200014062.mm.1 NM_007952.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014062.mm.1 NM_007952.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200014062.mm.1 TC0200001812.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0