This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00432880_mH
See other DNMT3A GE Assays ›
Gene Symbol:
Dnmt3a
Gene Name:
DNA methyltransferase 3A
Gene Aliases:
MmuIIIA
Chromosome Location:
Chr.12: 3804535 - 3914443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00432880_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnmt3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271753.1 NP_001258682.1 16 - 17 2187 63
NM_007872.4 NP_031898.1 16 - 17 2233 63
NM_153743.4 NP_714965.1 11 - 12 1411 63
GenBank mRNA AF068625.2 - 16 - 17 2137 63
AF480164.1 - 11 - 12 1411 63
AK077792.1 - 11 - 12 1376 63
AK090132.1 - 16 - 17 2164 63
AK147263.1 - 16 - 17 2187 63
AK147627.1 - 16 - 17 2187 63
AK147642.1 - 16 - 17 2187 63
AK147676.1 - 16 - 17 2162 63
AK157792.1 - 11 - 12 1382 63
BC007466.1 - 16 - 17 2165 3492853 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000016.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnmt3a

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA methylation
methylation-dependent chromatin silencing
regulation of gene expression by genetic imprinting
transcription, DNA-templated
regulation of transcription, DNA-templated
spermatogenesis
maintenance of DNA methylation
DNA methylation on cytosine within a CG sequence
positive regulation of cell death
chromatin modification
methylation
DNA methylation on cytosine
DNA methylation involved in embryo development
DNA methylation involved in gamete generation
hypermethylation of CpG island
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
cellular response to amino acid stimulus
C-5 methylation of cytosine
DNA binding
chromatin binding
DNA (cytosine-5-)-methyltransferase activity
protein binding
methyltransferase activity
transferase activity
identical protein binding
unmethylated CpG binding
metal ion binding
DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200000241.mm.1 NM_001271753.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000241.mm.1 NM_007872.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000241.mm.1 NM_007872.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200000241.mm.1 NM_153743.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200000241.mm.1 TC1200000016.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0