This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00432750_g1
See other CSK GE Assays ›
Gene Symbol:
Csk
Gene Name:
c-src tyrosine kinase
Gene Aliases:
AW212630, p50CSK
Chromosome Location:
Chr.9: 57626646 - 57653626 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00432750_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Csk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK136876.1 - 4 - 5 960 61
AK137297.1 - 4 - 5 374 61
AK156058.1 - 4 - 5 512 61
AK170815.1 - 4 - 5 528 61
BC018394.1 - 4 - 5 487 4206879 61
BC052006.1 - 4 - 5 545 5685297 61
BC095962.1 - 6 - 7 868 61
U05247.1 - 2 - 3 239 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002377.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900019923.mm.1 NM_007783.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900019923.mm.1 TC0900002377.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0