This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00432530_m1
See other CHUK GE Assays ›
Gene Symbol:
Chuk
Gene Name:
conserved helix-loop-helix ubiquitous kinase
Gene Aliases:
AI256658, Chuk1, Fbx24, Fbxo24, IKBKA, IKK1, Ikka, NFKBIKA
Chromosome Location:
Chr.19: 44073334 - 44107506 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00432530_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chuk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162410.1 NP_001155882.1 2 - 3 290 65
NM_007700.2 NP_031726.2 2 - 3 290 65
GenBank mRNA AK018671.1 - 2 - 3 269 65
AK035326.1 - 2 - 3 247 65
AK150039.1 - 2 - 3 269 65
AK155393.1 - 2 - 3 291 65
AK164236.1 - 2 - 3 291 65
BC018243.1 - 2 - 3 282 4511275 65
U12473.1 - 2 - 3 258 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001552.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chuk

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelial sheet
protein phosphorylation
I-kappaB phosphorylation
Rho protein signal transduction
lactation
response to organic substance
phosphorylation
signal transduction by protein phosphorylation
osteoclast differentiation
response to lipopolysaccharide
mammary gland epithelial cell proliferation
response to muscle stretch
odontogenesis of dentin-containing tooth
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of transcription from RNA polymerase II promoter
striated muscle cell differentiation
mammary gland alveolus development
cellular response to tumor necrosis factor
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
IkappaB kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein homodimerization activity
protein heterodimerization activity
scaffold protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0