This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00431673_m1
See other ADCYAP1R1 GE Assays ›
Gene Symbol:
Adcyap1r1
Gene Name:
adenylate cyclase activating polypeptide 1 receptor 1
Gene Aliases:
2900024I10Rik, AI846590, PAC1, PAC1R, PACAP1-R
Chromosome Location:
Chr.6: 55451501 - 55501455 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00431673_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adcyap1r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025372.2 NP_001020543.1 6 - 7 795 81
NM_007407.4 NP_031433.3 6 - 7 795 81
GenBank mRNA AK042820.1 - 6 - 7 787 81
AK047304.1 - 5 - 6 298 81
AK081121.1 - 5 - 6 298 81
AK083108.1 - 6 - 7 793 81
AK147291.1 - 6 - 7 746 81
AK147609.1 - 6 - 7 536 81
BC067039.1 - 6 - 7 728 6852762 81
D82935.1 - 6 - 7 478 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000667.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adcyap1r1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
multicellular organism development
spermatogenesis
positive regulation of calcium ion transport into cytosol
cAMP-mediated signaling
cell differentiation
positive regulation of cAMP biosynthetic process
negative regulation of cell death
positive regulation of inositol phosphate biosynthetic process
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
vasoactive intestinal polypeptide receptor activity
protein binding
adenylate cyclase binding
ADP-ribosylation factor binding
neuropeptide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005166.mm.1 NM_001025372.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600005166.mm.1 NM_007407.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600005166.mm.1 TC0600000667.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0