Assay Details
Gene Symbol: CARD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015435648.1 | XP_015291134.1 | 2 - 3 | 806 | 117 | |
XM_015435649.1 | XP_015291135.1 | 2 - 3 | 726 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
