This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04946705_m1
See other APOC2 GE Assays ›
Gene Symbol:
APOC2
Gene Name:
apolipoprotein C2
Gene Aliases:
Apo-CII, ApoC-II
Chromosome Location:
Chr.Unknown: 60328 - 69356 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04946705_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284711.1 NP_001271640.1 316 70
XM_015444652.1 XP_015300138.1 707 70
GenBank mRNA AK240618.1 - 128 70
M86345.1 - 316 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70

Panther Classification:

Molecular Function -

apolipoprotein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0