Assay Details
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542940.2 | XP_005542997.1 | 5 - 6 | 1066 | 99 | |
XM_015433730.1 | XP_015289216.1 | 6 - 7 | 1134 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
