Assay Details
Gene Symbol: SUMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547654.1 | XP_005547711.1 | 1 - 2 | 311 | 104 | |
XM_015446205.1 | XP_015301691.1 | 1 - 2 | 311 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
