This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04377487_m1
See other MGAT4D GE Assays ›
Gene Symbol:
MGAT4D
Gene Name:
MGAT4 family member D
Gene Aliases:
-
Chromosome Location:
Chr.5: 139794375 - 139847770 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04377487_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGAT4D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005555964.2 XP_005556021.1 6 - 7 1737 100
XM_015450852.1 XP_015306338.1 6 - 7 1737 100
XM_015450854.1 XP_015306340.1 6 - 7 1647 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0