Assay Details
Gene Symbol: C10H22orf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567328.2 | XP_005567385.1 | 3 - 4 | 282 | 66 | |
XM_005567329.2 | XP_005567386.1 | 3 - 4 | 280 | 66 | ||
XM_005567330.2 | XP_005567387.1 | 3 - 4 | 300 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
