This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04366661_g1
See other LOC102126770 GE Assays ›
Gene Symbol:
LOC102126770
Gene Name:
sarcoma antigen 1
Gene Aliases:
SAGE1
Chromosome Location:
Chr.X: 132624987 - 132640913 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04366661_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102126770
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005594666.2 XP_005594723.1 11 - 12 1881 122
XM_005594667.2 XP_005594724.1 11 - 12 1876 122
XM_005594669.2 XP_005594726.1 11 - 12 1867 122
XM_005594670.2 XP_005594727.1 10 - 11 1741 122
XM_005594671.2 XP_005594728.1 11 - 12 1726 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

RNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0