This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04362080_m1
See other LIMS2 GE Assays ›
Gene Symbol:
LIMS2
Gene Name:
LIM zinc finger domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 3943203 - 3987360 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04362080_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283510.1 NP_001270439.1 4 - 5 420 66
XM_005572857.2 XP_005572914.1 4 - 5 66
XM_005572859.2 XP_005572916.1 4 - 5 66
XM_005572860.2 XP_005572917.1 4 - 5 66
XM_005572863.2 XP_005572920.1 5 - 6 66
XM_015431914.1 XP_015287400.1 3 - 4 66
GenBank mRNA AB174615.1 - 4 - 5 420 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

cell junction protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0