This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04356844_m1
See other DYRK3 GE Assays ›
Gene Symbol:
DYRK3
Gene Name:
dual specificity tyrosine phosphorylation regulated kinase 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 65203490 - 65218154 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04356844_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DYRK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285247.1 NP_001272176.1 3 - 4 279 89
XM_005540643.2 XP_005540700.1 2 - 3 360 89
XM_005540645.2 XP_005540702.1 3 - 4 422 89
XM_015455792.1 XP_015311278.1 2 - 3 328 89
GenBank mRNA AB169105.1 - 3 - 4 280 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0