This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04356206_m1
See other CEP43 GE Assays ›
Gene Symbol:
CEP43
Gene Name:
centrosomal protein 43
Gene Aliases:
FGFR1OP
Chromosome Location:
Chr.4: 3680272 - 3725653 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04356206_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEP43
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283512.1 NP_001270441.1 11 - 12 1084 94
XM_005551444.2 XP_005551501.1 12 - 13 1919 94
XM_005551445.2 XP_005551502.1 12 - 13 1916 94
XM_005551448.2 XP_005551505.1 11 - 12 1856 94
GenBank mRNA AB168708.1 - 11 - 12 1084 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0