This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04355692_m1
See other DNAJB4 GE Assays ›
Gene Symbol:
DNAJB4
Gene Name:
DnaJ heat shock protein family (Hsp40) member B4
Gene Aliases:
-
Chromosome Location:
Chr.1: 149205050 - 149243705 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04355692_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNAJB4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287310.1 NP_001274239.1 2 - 3 970 145
XM_005542952.2 XP_005543009.1 3 - 4 1112 145
XM_005542953.2 XP_005543010.1 3 - 4 827 145
XM_015452345.1 XP_015307831.1 3 - 4 1057 145
XM_015452362.1 XP_015307848.1 2 - 3 1023 145
XM_015452363.1 XP_015307849.1 2 - 3 583 145
GenBank mRNA AB173150.1 - 2 - 3 970 145

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0