This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04346799_m1
See other S100PBP GE Assays ›
Gene Symbol:
S100PBP
Gene Name:
S100P binding protein
Gene Aliases:
-
Chromosome Location:
Chr.1: 195101390 - 195145341 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04346799_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100PBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005544076.2 XP_005544133.1 4 - 5 1215 101
XM_005544077.2 XP_005544134.1 4 - 5 1148 101
XM_005544078.2 XP_005544135.1 3 - 4 978 101
XM_005544079.2 XP_005544136.1 4 - 5 1150 101
XM_015438965.1 XP_015294451.1 4 - 5 1155 101
XM_015438978.1 XP_015294464.1 4 - 5 1153 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0