This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04253421_m1
See other GDPGP1 GE Assays ›
Gene Symbol:
GDPGP1
Gene Name:
GDP-D-glucose phosphorylase 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 70544753 - 70553464 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04253421_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDPGP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319429.1 NP_001306358.1 3 - 4 184 92
XM_015453395.1 XP_015308881.1 2 - 3 392 92
XM_015453396.1 XP_015308882.1 2 - 3 86 92
GenBank mRNA AB093646.1 - 3 - 4 184 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001319429.1, XM_015453395.1, XM_015453396.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0